Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042486.3 | NP_001035951.1 | 2 | 159 | ||
NM_014902.5 | NP_055717.2 | 8 | 2204 | |||
NM_183006.3 | NP_892118.1 | 2 | 880 | |||
XM_005260332.4 | XP_005260389.1 | 2 | 106 | |||
XM_005260333.4 | XP_005260390.1 | 2 | 106 | |||
XM_011528688.2 | XP_011526990.1 | 7 | 1970 | |||
XM_011528689.2 | XP_011526991.1 | 7 | 1977 | |||
XM_011528691.2 | XP_011526993.1 | 3 | 1019 | |||
XM_011528692.2 | XP_011526994.1 | 3 | 209 | |||
XM_017027725.1 | XP_016883214.1 | 2 | 916 | |||
XM_017027726.1 | XP_016883215.1 | 3 | 205 | |||
XM_017027727.1 | XP_016883216.1 | 2 | 135 | |||
GenBank mRNA | AB023181.1 | - | 8 | 2146 | ||
AK001681.1 | - | 2 | 106 | |||
AK125383.1 | - | 2 | 866 | |||
BC023214.1 | - | 2 | 277 | |||
BC108706.1 | - | 2 | 869 | |||
BC153874.1 | - | 2 | 880 | |||
CN419826.1 | - | 1 | 23 | |||
DA688888.1 | - | 2 | 108 | |||
DB283453.1 | - | 2 | 136 | |||
HY068872.1 | - | 2 | 161 |