Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002881.2 | NP_002872.1 | 3 | 460 | ||
XM_005263724.1 | XP_005263781.1 | 3 | 373 | |||
XM_005263727.1 | XP_005263784.1 | 3 | 462 | |||
XM_005263728.1 | XP_005263785.1 | 3 | 667 | |||
XM_005263729.2 | XP_005263786.1 | 3 | 718 | |||
XM_011511573.1 | XP_011509875.1 | 3 | 370 | |||
XM_011511574.1 | XP_011509876.1 | 4 | 521 | |||
XM_017004621.1 | XP_016860110.1 | 3 | 411 | |||
XM_017004622.1 | XP_016860111.1 | 4 | 559 | |||
GenBank mRNA | AF493911.1 | - | 2 | 270 | ||
AK127675.1 | - | 4 | 508 | |||
AK303214.1 | - | 3 | 379 | |||
AK312453.1 | - | 2 | 284 | |||
BC018163.1 | - | 3 | 365 | 3880116 | ||
BQ893549.1 | - | 465 | ||||
BT006953.1 | - | 2 | 270 | |||
M35416.1 | - | 3 | 440 | |||
X15015.1 | - | 2 | 270 |