Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015557.2 | NP_056372.1 | 3 | 331 | ||
GenBank mRNA | AF425231.1 | - | 3 | 320 | ||
AK127046.1 | - | 3 | 220 |