Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039667.2 | NP_001034756.1 | 6 | 1340 | ||
NM_139314.2 | NP_647475.1 | 7 | 1454 | |||
XM_005272484.3 | XP_005272541.1 | 7 | 1506 | |||
XM_005272485.3 | XP_005272542.1 | 6 | 1392 | |||
GenBank mRNA | AB056477.1 | - | 7 | 1428 | ||
AF153606.1 | - | 7 | 1409 | |||
AF169312.1 | - | 7 | 1323 | |||
AF202636.1 | - | 7 | 1431 | |||
AK222489.1 | - | 7 | 1426 | |||
AK303269.1 | - | 8 | 1355 | |||
AW954426.1 | - | 1 | 145 | |||
AY358275.1 | - | 7 | 1422 | |||
BC023647.2 | - | 7 | 1398 | 5088323 |