Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017580.2 | NP_060050.2 | 9 | 2535 | ||
XM_005269925.4 | XP_005269982.1 | 10 | 2390 | |||
XM_005269926.4 | XP_005269983.1 | 10 | 2578 | |||
XM_006717907.2 | XP_006717970.1 | 10 | 3260 | |||
XM_011539887.2 | XP_011538189.1 | 5 | 821 | |||
XM_017016357.1 | XP_016871846.1 | 10 | 3991 | |||
XM_017016358.1 | XP_016871847.1 | 10 | 2225 | |||
GenBank mRNA | AJ252060.1 | - | 9 | 2570 | ||
AK302810.1 | - | 10 | 2346 | |||
AL832925.1 | - | 9 | 1715 |