Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145648.1 | NP_001139120.1 | 24 | 3842 | ||
NM_002891.4 | NP_002882.3 | 25 | 3890 | |||
NM_153815.2 | NP_722522.1 | 11 | 2336 | |||
XM_011521866.2 | XP_011520168.1 | 25 | 3790 | |||
XM_011521867.2 | XP_011520169.1 | 25 | 3790 | |||
XM_017022455.1 | XP_016877944.1 | 25 | 3781 | |||
XM_017022456.1 | XP_016877945.1 | 24 | 3751 | |||
XM_017022457.1 | XP_016877946.1 | 24 | 3751 | |||
XM_017022458.1 | XP_016877947.1 | 24 | 3742 | |||
XM_017022459.1 | XP_016877948.1 | 24 | 3751 | |||
GenBank mRNA | AK226101.1 | - | 23 | 3126 | ||
AK309047.1 | - | 23 | 3273 | |||
AL359931.1 | - | 1 | 1020 | |||
L26584.1 | - | 25 | 3521 | |||
M91815.1 | - | 4 | 383 | |||
S62035.1 | - | 11 | 2345 |