Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304746.1 | NP_001291675.1 | 5 | 838 | ||
NM_017821.4 | NP_060291.2 | 5 | 868 | |||
GenBank mRNA | AK097621.1 | - | 5 | 1603 | ||
AK294115.1 | - | 5 | 838 | |||
AY126343.1 | - | 4 | 594 | |||
BC137108.1 | - | 4 | 717 | 9021105 | ||
BC137110.1 | - | 4 | 717 | 9021107 | ||
BC144476.1 | - | 4 | 717 |