Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310332.1 | NP_001297261.1 | 7 | 986 | ||
NM_017999.4 | NP_060469.4 | 7 | 1330 | |||
GenBank mRNA | AB265810.1 | - | 7 | 1081 | ||
AK055542.1 | - | 1 | 127 | |||
AK122711.1 | - | 6 | 783 | |||
AK225832.1 | - | 7 | 1280 | |||
AK291247.1 | - | 7 | 1282 | |||
AY256461.1 | - | 7 | 974 | |||
BC012077.1 | - | 7 | 1250 | 4653017 |