Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310332.1 | NP_001297261.1 | 21 | 3193 | ||
NM_017999.4 | NP_060469.4 | 21 | 3537 | |||
GenBank mRNA | AK000973.1 | - | 14 | 1786 | ||
AK027154.1 | - | 13 | 1933 | |||
AK055542.1 | - | 15 | 2334 | |||
AK122711.1 | - | 20 | 2822 | |||
AK225832.1 | - | 21 | 3487 | |||
AK291247.1 | - | 21 | 3489 | |||
AY256461.1 | - | 21 | 3181 | |||
BC009821.2 | - | 14 | 1891 | |||
BC012077.1 | - | 21 | 3457 | 4653017 | ||
BC017376.2 | - | 14 | 1881 |