Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HHAT SIRNA Assays ›
Gene Symbol:
HHAT
Gene Name:
hedgehog acyltransferase
Gene Aliases:
MART2, SKI1, Skn
Chromosome Location:
Chr.1: 210327873 - 210676298 on Build GRCh38
UniGene ID:
Hs.58650
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121827
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HHAT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122834.3
NP_001116306.1
4
351
NM_001170564.2
NP_001164035.1
4
351
NM_001170580.2
NP_001164051.1
4
406
NM_001170587.2
NP_001164058.1
3
283
NM_001170588.2
NP_001164059.1
4
417
NM_018194.5
NP_060664.2
4
417
XM_006711441.3
XP_006711504.1
5
478
XM_011509737.2
XP_011508039.1
5
478
XM_011509738.2
XP_011508040.1
3
236
XM_011509739.2
XP_011508041.1
4
403
XM_011509740.2
XP_011508042.1
5
607
XM_011509741.2
XP_011508043.1
4
351
XM_011509742.2
XP_011508044.1
5
455
XM_011509744.2
XP_011508046.1
3
411
XM_011509746.2
XP_011508048.1
4
432
XM_011509747.1
XP_011508049.1
2
201
XM_017001726.1
XP_016857215.1
3
452
XM_017001727.1
XP_016857216.1
3
452
XM_017001728.1
XP_016857217.1
4
561
XM_017001729.1
XP_016857218.1
4
282
XM_017001730.1
XP_016857219.1
3
364
XM_017001731.1
XP_016857220.1
3
170
XM_017001732.1
XP_016857221.1
4
242
XM_017001733.1
XP_016857222.1
2
324
XM_017001734.1
XP_016857223.1
3
358
XM_017001735.1
XP_016857224.1
4
882
XM_017001736.1
XP_016857225.1
3
258
XM_017001737.1
XP_016857226.1
4
242
XM_017001738.1
XP_016857227.1
4
349
XM_017001739.1
XP_016857228.1
4
438
XM_017001740.1
XP_016857229.1
4
939
XM_017001741.1
XP_016857230.1
2
100
XM_017001742.1
XP_016857231.1
2
739
XM_017001743.1
XP_016857232.1
2
96
XM_017001744.1
XP_016857233.1
2
528
GenBank mRNA
AK001586.1
-
4
371
AK295130.1
-
3
303
AK297193.1
-
4
349
AK298991.1
-
4
371
AK299200.1
-
2
201
AK302955.1
-
3
236
AK316190.1
-
3
237
AK316524.1
-
4
464
AL049848.1
-
2
84
BC039071.1
-
4
359
5177926
BC117130.1
-
3
206
40125681
CR936628.1
-
2
85
DA336461.1
-
4
403
DB077466.1
-
4
341
DB080645.1
-
4
351
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
smoothened signaling pathway
multicellular organism development
protein palmitoylation
positive regulation of protein targeting to mitochondrion
GTP binding
O-acyltransferase activity
palmitoyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline