Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018218.2 | NP_060688.1 | 2 | 293 | ||
XM_006712612.3 | XP_006712675.1 | 2 | 308 | |||
XM_011511396.2 | XP_011509698.1 | 2 | 308 | |||
XM_011511397.2 | XP_011509699.1 | 3 | 382 | |||
XM_011511398.2 | XP_011509700.1 | 2 | 308 | |||
XM_011511399.2 | XP_011509701.1 | 3 | 382 | |||
XM_011511400.2 | XP_011509702.1 | 1 | 93 | |||
XM_011511401.2 | XP_011509703.1 | 2 | 308 | |||
XM_017004426.1 | XP_016859915.1 | 3 | 382 | |||
XM_017004428.1 | XP_016859917.1 | 2 | 192 | |||
GenBank mRNA | AJ583821.2 | - | 2 | 293 | ||
AK001647.1 | - | 2 | 255 | |||
AK295112.1 | - | 3 | 375 | |||
BC067300.1 | - | 3 | 355 |