Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 6 | 502 | ||
NM_001271015.1 | NP_001257944.1 | 5 | 469 | |||
NM_018299.4 | NP_060769.4 | 5 | 469 | |||
NR_073119.1 | 4 | 366 | ||||
NR_073120.1 | 4 | 366 | ||||
NR_073121.1 | 4 | 366 | ||||
GenBank mRNA | AK001873.1 | - | 5 | 384 | ||
AK021735.1 | - | 5 | 384 | |||
AK024050.1 | - | 6 | 467 | |||
AK225554.1 | - | 5 | 400 | |||
AK295792.1 | - | 5 | 469 | |||
AY948289.1 | - | 6 | 438 | |||
BC010900.1 | - | 5 | 388 | 4081461 | ||
BC016326.1 | - | 5 | 423 | 4093131 | ||
BU155172.1 | - | 4 | 312 | |||
BX538145.1 | - | 4 | 289 | |||
BX648427.1 | - | 4 | 295 | |||
CR457275.1 | - | 4 | 381 | |||
DA352184.1 | - | 4 | 319 |