Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199198.2 | NP_001186127.1 | 2 | 298 | ||
NM_018309.4 | NP_060779.2 | 2 | 298 | |||
XM_011512974.2 | XP_011511276.1 | 1 | 43 | |||
XM_017006840.1 | XP_016862329.1 | 2 | 130 | |||
XM_017006841.1 | XP_016862330.1 | 2 | 115 | |||
GenBank mRNA | AB449910.1 | - | 2 | 96 | ||
AK001908.1 | - | 2 | 103 | |||
BC020955.1 | - | 2 | 92 | 3847561 | ||
BX648032.1 | - | 2 | 90 | |||
DA801258.1 | - | 2 | 298 | |||
DB458401.1 | - | 2 | 273 |