Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018362.3 | NP_060832.1 | 5 | 533 | ||
GenBank mRNA | AK002077.1 | - | 5 | 527 | ||
AL833601.1 | - | 4 | 452 | |||
BC009774.1 | - | 5 | 520 | |||
BC053907.1 | - | 5 | 510 | 6502685 | ||
DA231006.1 | - | 5 | 533 |