Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040450.2 | NP_001035540.1 | 7 | 1655 | ||
NM_001040453.2 | NP_001035543.1 | 7 | 1485 | |||
XM_011521687.1 | XP_011519989.1 | 8 | 1707 | |||
XM_011521688.1 | XP_011519990.1 | 8 | 1707 | |||
XM_011521689.1 | XP_011519991.1 | 8 | 1605 | |||
XM_011521690.1 | XP_011519992.1 | 7 | 1551 | |||
XM_017022332.1 | XP_016877821.1 | 6 | 1461 | |||
XM_017022333.1 | XP_016877822.1 | 5 | 1407 | |||
XM_017022334.1 | XP_016877823.1 | 5 | 1407 | |||
GenBank mRNA | AB032990.1 | - | 7 | 762 | ||
AK075319.1 | - | 7 | 1533 | |||
AK297291.1 | - | 7 | 1533 | |||
AL833610.1 | - | 6 | 1513 | |||
BC140811.1 | - | 7 | 1485 | 9021679 | ||
BC144616.1 | - | 7 | 1485 | 9053147 |