Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020135.2 | NP_064520.2 | 7 | 2221 | ||
NM_130395.2 | NP_569079.1 | 7 | 2146 | |||
GenBank mRNA | AB056152.1 | - | 7 | 2222 | ||
AB209723.1 | - | 7 | 1974 | |||
AF218313.1 | - | 7 | 1608 | |||
AK026179.1 | - | 6 | 1126 | |||
AK094823.1 | - | 6 | 2028 | |||
AK223593.1 | - | 7 | 2155 | |||
BC018923.2 | - | 7 | 2107 | 4109718 |