Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020529.2 | NP_065390.1 | 6 | 1267 | ||
GenBank mRNA | AK311515.1 | - | 5 | 1029 | ||
BC002601.2 | - | 6 | 1261 | 3161686 | ||
BC004983.1 | - | 6 | 1250 | 2957970 | ||
M69043.1 | - | 6 | 1251 |