Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021146.3 | NP_066969.1 | 1 | 392 | ||
XM_017000004.1 | XP_016855493.1 | 1 | 357 | |||
GenBank mRNA | AB593026.1 | - | 1 | 326 | ||
AB593027.1 | - | 1 | 393 | |||
AK313716.1 | - | 1 | 358 | |||
AY358301.1 | - | 1 | 315 | |||
BC001881.1 | - | 1 | 340 | 3544149 | ||
BT009802.1 | - | 1 | 101 | |||
CA391009.1 | - | 1 | 392 | |||
Y16132.1 | - | 1 | 108 |