Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000509.5 | NP_000500.2 | 4 | 577 | ||
NM_021870.2 | NP_068656.2 | 4 | 519 | |||
GenBank mRNA | AK289422.1 | - | 4 | 422 | ||
AK290824.1 | - | 4 | 468 | |||
BC007044.1 | - | 4 | 424 | 3934943 | ||
BC021674.1 | - | 4 | 426 | 3935073 | ||
BG567044.1 | - | 4 | 519 | |||
BT007081.1 | - | 4 | 378 | |||
BX382942.2 | - | 4 | 617 | |||
BX538098.1 | - | 3 | 266 | |||
X51473.1 | - | 2 | 156 |