Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287516.1 | NP_001274445.1 | 16 | 2475 | ||
NM_001287517.1 | NP_001274446.1 | 16 | 2433 | |||
NM_001287518.1 | NP_001274447.1 | 15 | 2394 | |||
NM_001287519.1 | NP_001274448.1 | 16 | 2206 | |||
NM_001287520.1 | NP_001274449.1 | 16 | 2248 | |||
NM_001287522.1 | NP_001274451.1 | 15 | 2125 | |||
NM_001287524.1 | NP_001274453.1 | 12 | 2030 | |||
NM_004358.4 | NP_004349.1 | 16 | 3058 | |||
NM_021872.3 | NP_068658.1 | 15 | 2977 | |||
NM_021873.3 | NP_068659.1 | 16 | 3100 | |||
NR_136335.1 | 17 | 2327 | ||||
NR_136336.1 | 17 | 2554 | ||||
GenBank mRNA | AK124284.1 | - | 1 | 1464 | ||
AL561439.3 | - | 1 | 94 | |||
BC006395.2 | - | 16 | 2381 | 4135465 | ||
BC009953.2 | - | 16 | 2461 | 4130804 | ||
BC051711.1 | - | 16 | 3005 | 5301205 | ||
BM793120.1 | - | 1 | 296 | |||
BX640836.1 | - | 12 | 2023 | |||
FR695900.1 | - | 16 | 2248 | |||
FR695901.1 | - | 15 | 2125 | |||
M81934.1 | - | 16 | 2335 | |||
S78187.1 | - | 16 | 2503 |