Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100812.1 | NP_001094282.1 | 3 | 881 | ||
NM_022059.3 | NP_071342.2 | 3 | 881 | |||
GenBank mRNA | AF275260.1 | - | 3 | 392 | ||
AF301016.1 | - | 3 | 772 | |||
AF337812.1 | - | 3 | 306 | |||
AK027389.1 | - | 3 | 799 | |||
AK292114.1 | - | 3 | 723 | |||
AK315017.1 | - | 3 | 881 | |||
AY358909.1 | - | 3 | 726 | |||
BC017588.1 | - | 3 | 875 | 4837010 | ||
BC044930.1 | - | 3 | 576 | 5762458 | ||
JC209418.1 | - | 3 | 881 | |||
JC209420.1 | - | 3 | 881 |