Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030761.4 | NP_110388.2 | 5 | 1483 | ||
XM_011541597.2 | XP_011539899.1 | 5 | 2957 | |||
XM_011541598.2 | XP_011539900.1 | 5 | 1552 | |||
GenBank mRNA | AY358947.1 | - | 5 | 1415 |