Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032873.4 | NP_116262.2 | 10 | 1744 | ||
XM_005271712.3 | XP_005271769.1 | 10 | 3990 | |||
XM_011543041.2 | XP_011541343.1 | 10 | 1382 | |||
XM_011543042.1 | XP_011541344.1 | 10 | 1748 | |||
GenBank mRNA | AB075839.1 | - | 10 | 1596 | ||
AF425252.1 | - | 8 | 1041 | |||
AK075203.1 | - | 10 | 1712 | |||
BC007541.1 | - | 10 | 1698 | 2958242 |