Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002916.3 | NP_002907.1 | 3, 4 | 433 | ||
NM_181573.2 | NP_853551.1 | 3, 4 | 492 | |||
GenBank mRNA | AK297282.1 | - | 3, 4 | 400 | ||
BC017452.1 | - | 3, 4 | 370 | 3537752 | ||
BC024022.1 | - | 3, 4 | 386 | 4658221 | ||
BM837975.1 | - | 3, 4 | 298 | |||
BT006987.1 | - | 2, 3 | 210 | |||
CR536561.1 | - | 2, 3 | 210 | |||
CX788139.1 | - | 3, 4 | 492 | |||
DA359927.1 | - | 3, 4 | 433 |