Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133367.4 | NP_588608.1 | 2 | 1477 | ||
GenBank mRNA | AF313619.1 | - | 1 | 909 | ||
AF347029.1 | - | 2 | 1467 | |||
AK093603.1 | - | 1 | 1093 | |||
AK128085.1 | - | 1 | 1697 | |||
AY424286.1 | - | 3 | 1480 | |||
BC030664.2 | - | 2 | 1452 | 5260230 |
Set Membership: |
G-protein coupled receptor |