Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002922.3 | NP_002913.3 | 1, 2 | 196 | ||
GenBank mRNA | AF493925.1 | - | 1, 2 | 91 | ||
AK093544.1 | - | 1, 2 | 148 | |||
AK303138.1 | - | 1, 2 | 150 | |||
AK315607.1 | - | 1, 2 | 150 | |||
AL039059.1 | - | 1, 2 | 200 | |||
BC015510.1 | - | 1, 2 | 128 | 3916789 | ||
BG536972.1 | - | 1, 2 | 196 | |||
BT006668.1 | - | 1, 2 | 91 | |||
EU668340.1 | - | 1, 2 | 128 | |||
X73427.1 | - | 1, 2 | 105 |