Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138328.2 | NP_612201.1 | 6 | 765 | ||
XM_006721733.3 | XP_006721796.1 | 5 | 742 | |||
XM_006721734.3 | XP_006721797.1 | 5 | 1134 | |||
XM_011524443.2 | XP_011522745.1 | 4 | 583 | |||
XM_017024272.1 | XP_016879761.1 | 6 | 770 | |||
XM_017024273.1 | XP_016879762.1 | 5 | 745 | |||
XM_017024274.1 | XP_016879763.1 | 5 | 1073 | |||
XM_017024275.1 | XP_016879764.1 | 5 | 611 | |||
XM_017024276.1 | XP_016879765.1 | 4 | 585 | |||
XM_017024277.1 | XP_016879766.1 | 5 | 674 | |||
XM_017024278.1 | XP_016879767.1 | 5 | 701 | |||
XM_017024279.1 | XP_016879768.1 | 4 | 1408 | |||
XM_017024280.1 | XP_016879769.1 | 3 | 725 | |||
GenBank mRNA | AJ313480.1 | - | 6 | 765 | ||
AK124570.1 | - | 4 | 579 | |||
BC100975.1 | - | 6 | 764 | 40005244 | ||
BC100976.1 | - | 6 | 765 | |||
BC100977.1 | - | 5 | 741 | 40005246 | ||
BC100978.1 | - | 6 | 744 | 40005247 |