Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005339.1 | NP_001005339.1 | 2 | 231 | ||
NM_002925.3 | NP_002916.1 | 2 | 180 | |||
XM_017016492.1 | XP_016871981.1 | 2 | 228 | |||
XM_017016493.1 | XP_016871982.1 | 2 | 273 | |||
GenBank mRNA | AF045229.2 | - | 2 | 160 | ||
AF368902.1 | - | 1 | 135 | |||
AF493934.1 | - | 1 | 117 | |||
AK290773.1 | - | 2 | 186 | |||
AL547312.3 | - | 2 | 180 | |||
AV707839.1 | - | 2 | 180 | |||
BC009361.2 | - | 2 | 176 | 4301597 | ||
BF685740.1 | - | 2 | 177 | |||
BG763741.1 | - | 2 | 187 | |||
BM914660.1 | - | 230 | ||||
CB124184.1 | - | 2 | 231 | |||
CR457008.1 | - | 1 | 117 |