Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139246.4 | NP_640339.4 | 3 | 961 | ||
XM_011518312.2 | XP_011516614.1 | 2 | 424 | |||
GenBank mRNA | AF258575.1 | - | 2 | 407 | ||
AK056453.1 | - | 1 | 88 | |||
AK094043.1 | - | 3 | 674 | |||
AK122809.1 | - | 1 | 1075 | |||
AK128615.1 | - | 4 | 627 | |||
AK291199.1 | - | 3 | 582 | |||
BC035604.1 | - | 3 | 608 | 4385905 | ||
BX538308.1 | - | 3 | 602 | |||
BX648493.1 | - | 3 | 600 | |||
KU178877.1 | - | 2 | 392 | |||
KU178878.1 | - | 2 | 364 |