Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002928.3 | NP_002919.3 | 4 | 459 | ||
GenBank mRNA | AF493937.1 | - | 4 | 305 | ||
AK301261.1 | - | 4 | 435 | |||
AK311880.1 | - | 4 | 435 | |||
AW193671.1 | - | 4 | 454 | |||
BC006243.2 | - | 4 | 421 | 3946461 | ||
BT006638.1 | - | 4 | 305 | |||
DA060276.1 | - | 1 | 177 | |||
DC349953.1 | - | 4 | 460 | |||
U70426.1 | - | 4 | 397 | |||
U94829.1 | - | 4 | 308 |