Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025014.1 | NP_079290.1 | 16 | 3085 | ||
NM_173728.3 | NP_776089.2 | 16 | 3149 | |||
XM_011523734.1 | XP_011522036.1 | 16 | 3194 | |||
XM_011523735.1 | XP_011522037.1 | 16 | 3130 | |||
XM_011523736.1 | XP_011522038.1 | 16 | 3145 | |||
XM_011523737.1 | XP_011522039.1 | 13 | 2186 | |||
GenBank mRNA | AB020722.1 | - | 16 | 3065 | ||
BC036749.1 | - | 16 | 3104 | 5204409 |