Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFBKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135863.1 | NP_001129335.1 | 4 | 687 | ||
NM_001135864.1 | NP_001129336.1 | 4 | 668 | |||
NM_173802.3 | NP_776163.1 | 4 | 748 | |||
GenBank mRNA | AK290248.1 | - | 4 | 669 | ||
AL832339.1 | - | 4 | 674 | |||
BC039107.1 | - | 5 | 1139 | 4837913 | ||
BC039535.1 | - | 4 | 748 | 5744381 |