Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178130.3 | NP_835231.1 | 10 | 714 | ||
XM_005247429.3 | XP_005247486.1 | 10 | 695 | |||
XM_006713615.3 | XP_006713678.1 | 9 | 2577 | |||
XM_011512774.2 | XP_011511076.1 | 9 | 581 | |||
XM_017006306.1 | XP_016861795.1 | 10 | 698 | |||
XM_017006307.1 | XP_016861796.1 | 9 | 1147 | |||
XM_017006308.1 | XP_016861797.1 | 10 | 2694 | |||
XM_017006309.1 | XP_016861798.1 | 10 | 726 | |||
XM_017006310.1 | XP_016861799.1 | 9 | 656 | |||
XM_017006311.1 | XP_016861800.1 | 9 | 581 | |||
XM_017006312.1 | XP_016861801.1 | 9 | 2577 | |||
XM_017006313.1 | XP_016861802.1 | 11 | 3201 | |||
XM_017006314.1 | XP_016861803.1 | 10 | 1152 | |||
XM_017006315.1 | XP_016861804.1 | 8 | 630 | |||
XM_017006316.1 | XP_016861805.1 | 8 | 2505 | |||
XM_017006317.1 | XP_016861806.1 | 8 | 630 | |||
XM_017006318.1 | XP_016861807.1 | 7 | 958 | |||
XM_017006319.1 | XP_016861808.1 | 5 | 365 | |||
GenBank mRNA | AF196568.1 | - | 9 | 687 | ||
AK094830.1 | - | 10 | 714 | |||
AY336746.1 | - | 6 | 404 |