Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182519.2 | NP_872325.2 | 3 | 364 | ||
GenBank mRNA | AF549190.1 | - | 2 | 232 |