Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198584.2 | NP_940986.1 | 2 | 514 | ||
GenBank mRNA | AK095314.1 | - | 2 | 223 | ||
BC052602.1 | - | 2 | 474 | 6083105 | ||
DB287770.1 | - | 2 | 514 |