Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003013.2 | NP_003004.1 | 1, 2 | 747 | ||
GenBank mRNA | AF017986.1 | - | 1, 2 | 764 | ||
AF311912.1 | - | 1, 2 | 743 | |||
AK075372.1 | - | 1, 2 | 740 | |||
AK093922.1 | - | 2, 3 | 715 | |||
AY359001.1 | - | 1, 2 | 549 | |||
BC008666.1 | - | 1, 2 | 727 | 3874262 | ||
BM924815.1 | - | 1, 2 | 763 |
Set Membership: |
G-protein coupled receptor |