Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003013.2 | NP_003004.1 | 3 | 1292 | ||
GenBank mRNA | AF311912.1 | - | 3 | 1288 | ||
AK075372.1 | - | 3 | 1285 | |||
AK093922.1 | - | 4 | 1260 | |||
AY359001.1 | - | 3 | 1094 | |||
BC008666.1 | - | 3 | 1272 | 3874262 |
Set Membership: |
G-protein coupled receptor |