Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002167.4 | NP_002158.3 | 3 | 1033 | ||
GenBank mRNA | A17546.1 | - | 3 | 787 | ||
AK225965.1 | - | 1 | 1394 | |||
BC003107.1 | - | 3 | 751 | 3543936 | ||
X66924.1 | - | 3 | 787 | |||
X69111.1 | - | 3 | 1032 |