Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003396.2 | NP_003387.1 | 4 | 1394 | ||
XM_011525178.2 | XP_011523480.1 | 4 | 1504 | |||
GenBank mRNA | AB063483.1 | - | 4 | 1378 | ||
AK127268.1 | - | 4 | 1415 |