Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004626.2 | NP_004617.2 | 5 | 1025 | ||
XM_005274231.1 | XP_005274288.1 | 6 | 1281 | |||
XM_011545238.1 | XP_011543540.1 | 6 | 1311 | |||
XM_011545239.2 | XP_011543541.1 | 6 | 1519 | |||
XM_011545240.2 | XP_011543542.1 | 7 | 1475 | |||
XM_011545241.2 | XP_011543543.1 | 5 | 763 | |||
XM_017018241.1 | XP_016873730.1 | 4 | 1783 | |||
XM_017018242.1 | XP_016873731.1 | 4 | 1618 | |||
XM_017018243.1 | XP_016873732.1 | 3 | 437 | |||
GenBank mRNA | AB070218.1 | - | 5 | 964 | ||
AK313570.1 | - | 5 | 1027 | |||
BC074790.2 | - | 5 | 944 | 30915564 | ||
BC074791.2 | - | 5 | 944 | 30915347 | ||
BC113386.1 | - | 5 | 945 | 8322438 | ||
BC113388.1 | - | 5 | 945 | 8322440 | ||
BT019492.1 | - | 5 | 902 | |||
JF800675.1 | - | 5 | 626 | |||
JQ715612.1 | - | 5 | 781 | |||
JQ715613.1 | - | 5 | 590 |