Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005524.3 | NP_005515.1 | 1 | 334 | ||
GenBank mRNA | AF264785.1 | - | 1 | 95 | ||
AK000415.1 | - | 1 | 331 | |||
AK300482.1 | - | 1 | 325 | |||
BC039152.1 | - | 1 | 282 | 4749611 | ||
CR541843.1 | - | 1 | 95 |