Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002960.1 | NP_002951.1 | 3 | 298 | ||
GenBank mRNA | BC012893.1 | - | 3 | 282 | 3880514 | |
BT006955.1 | - | 2 | 215 | |||
CR542163.1 | - | 2 | 215 | |||
CR542185.1 | - | 2 | 215 | |||
Z18948.1 | - | 3 | 298 |