Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001900.4 | NP_001891.2 | 2, 3 | 405 | ||
GenBank mRNA | BC062678.1 | - | 2, 3 | 402 | 30349574 | |
BC069514.1 | - | 2, 3 | 342 | 7262733 | ||
CB985957.1 | - | 408 | ||||
X70377.1 | - | 2, 3 | 342 |