Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005185.3 | NP_005176.1 | 1 | 915 | ||
GenBank mRNA | BC031889.1 | - | 1 | 823 | 4777524 | |
CU454493.1 | - | 1 | 263 | |||
M36707.1 | - | 1 | 1783 | |||
M58026.1 | - | 1 | 843 |