Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MESP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018670.3 | NP_061140.1 | 2 | 1105 | ||
GenBank mRNA | AL357533.1 | - | 2 | 658 | ||
AL357534.1 | - | 2 | 636 | |||
AL357535.1 | - | 1 | 278 | |||
AL357570.1 | - | 2 | 552 | |||
BC006219.1 | - | 2 | 1113 | 3944350 |