Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019071.2 | NP_061944.2 | 4 | 379 | ||
NM_198267.1 | NP_938008.1 | 4 | 379 | |||
XM_017012369.1 | XP_016867858.1 | 4 | 387 | |||
GenBank mRNA | AF074968.1 | - | 4 | 356 | ||
AF161419.1 | - | 4 | 369 | |||
AF180298.1 | - | 4 | 346 | |||
AK000096.1 | - | 4 | 380 | |||
AK291905.1 | - | 4 | 357 | |||
AK293682.1 | - | 3 | 265 | |||
AL603623.1 | - | 4 | 356 | |||
AY007790.1 | - | 4 | 360 | |||
BC009776.1 | - | 4 | 334 | |||
BC009777.1 | - | 4 | 367 | |||
BC010851.1 | - | 4 | 326 | |||
BC062634.1 | - | 4 | 342 | |||
BC073865.1 | - | 4 | 347 | |||
BC093091.1 | - | 4 | 363 | |||
BC093689.1 | - | 4 | 339 | |||
BC101609.1 | - | 4 | 339 | |||
BC143316.1 | - | 4 | 339 |