Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323956.1 | NP_001310885.1 | 8 | 1686 | ||
NM_001323958.1 | NP_001310887.1 | 7 | 1541 | |||
NM_006462.5 | NP_006453.1 | 8 | 1702 | |||
NM_031229.3 | NP_112506.2 | 9 | 1847 | |||
NR_136659.1 | 9 | 1822 | ||||
XM_005260645.1 | XP_005260702.1 | 9 | 1643 | |||
XM_011529137.1 | XP_011527439.1 | 10 | 1766 | |||
XM_011529138.1 | XP_011527440.1 | 10 | 1721 | |||
XM_011529139.2 | XP_011527441.1 | 10 | 1766 | |||
XM_017027594.1 | XP_016883083.1 | 8 | 1731 | |||
XM_017027595.1 | XP_016883084.1 | 8 | 1076 | |||
GenBank mRNA | AB107766.1 | - | 8 | 1033 | ||
AK297524.1 | - | 8 | 1432 | |||
BC000983.2 | - | 7 | 1236 | |||
BC015219.2 | - | 9 | 1560 | 3877587 | ||
U67322.1 | - | 8 | 1588 |