Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181718.3 | NP_859069.2 | 1 | 993 | ||
XM_017023107.1 | XP_016878596.1 | 2 | 974 | |||
GenBank mRNA | AF070642.1 | - | 1 | 334 | ||
BC009033.1 | - | 1 | 720 | |||
BC029136.1 | - | 1 | 429 | |||
BC034275.3 | - | 1 | 466 | |||
BC040933.2 | - | 1 | 505 | |||
BC085011.1 | - | 1 | 841 | |||
BC126319.1 | - | 1 | 961 | 8992035 | ||
BC126321.1 | - | 1 | 961 | 8992037 | ||
BC144152.1 | - | 1 | 964 | 9052672 |