Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001675.4 | NP_001666.2 | 2 | 1901 | ||
NM_182810.2 | NP_877962.1 | 3 | 1294 | |||
XM_017028807.1 | XP_016884296.1 | 4 | 1773 | |||
GenBank mRNA | AK057751.1 | - | 3 | 1298 | ||
AK296028.1 | - | 2 | 1401 | |||
BC008090.1 | - | 3 | 1278 | 3454473 | ||
BC011994.1 | - | 3 | 1276 | 3882614 | ||
BC016855.2 | - | 3 | 1270 | 3850361 | ||
BC022088.2 | - | 3 | 1277 | 3030097 | ||
BC024775.1 | - | 3 | 1276 | 4992862 | ||
BC044895.1 | - | 3 | 1262 | 4513828 | ||
BC073754.1 | - | 3 | 1267 | 6580199 | ||
BC073990.1 | - | 3 | 1242 | 6022488 | ||
CR450353.1 | - | 2 | 1014 | |||
CR456384.1 | - | 2 | 1194 | |||
D90209.1 | - | 2 | 1895 | |||
M86842.1 | - | 2 | 1121 |