Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261400.2 | NP_001248329.1 | 1 | 227 | ||
NM_002813.6 | NP_002804.2 | 1 | 227 | |||
NR_048555.2 | 1 | 227 | ||||
GenBank mRNA | AK315389.1 | - | 1 | 196 | ||
BC002383.2 | - | 1 | 187 | 2961610 | ||
BC004184.2 | - | 1 | 187 | 2961610 | ||
BC004213.2 | - | 1 | 187 | 2961610 | ||
BI832046.1 | - | 1 | 221 | |||
BM850893.1 | - | 1 | 148 | |||
BU956605.1 | - | 1 | 208 | |||
CB111716.1 | - | 1 | 172 | |||
CN260241.1 | - | 1 | 233 |